ContactUsLinkComponent
Cynomolgus Mucin-1, His Tag MU1-C52H5 is expressed from human 293 cells HEK293. It contains AA Leu 254 - Gly 373 Accession XP_00554 1632.1. 1 * 1 mg
Catalog # ACRBMU1-C52H5-1MG
Supplier:  ACRO Biosystems
Cynomolgus Mucin-1, His Tag MU1-C52H5 is expressed from human 293 cells HEK293. It contains AA Leu 254 - Gly 373 Accession XP_00554 1632.1. 1 * 1 mg
Catalog # ACRBMU1-C52H5-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together