ContactUsLinkComponent
Human LIV-1, Fc Tag LV1-H5254 is expressed from human 293 cells HEK293. It contains AA Phe 29 - Trp 325 Accession Q13433-1. 1 * 100 µG
Catalog # ACRBLV1-H5254100UG
Supplier:  ACRO Biosystems
Human LIV-1, Fc Tag LV1-H5254 is expressed from human 293 cells HEK293. It contains AA Phe 29 - Trp 325 Accession Q13433-1. 1 * 100 µG
Catalog # ACRBLV1-H5254100UG
Supplier:  ACRO Biosystems
Frequently Bought Together