Anaerobic jar “eco” stainless steel/PC (transparent) ventilation screw, 3 litres capacity 120x270 mm (inner O x height) Suitable for up to 15 Petri dishes O 60-100 mm Optional racks: - for Petri dishes ( small , standard ) - for test tubes 1 * 1 ST
Artikelnr. SCUT3380502
Supplier: schuett-biotec GmbH
Frequently Bought Together