quick- disconnect adaptor M16x1 Temperature range: -45°C…220°C 1 * 1 items
Catalog # HUBE10787
Supplier: HUBER
quick- disconnect adaptor M16x1 Temperature range: -45°C…220°C 1 * 1 items
Catalog # HUBE10787
Supplier: HUBER
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
epT.I.P.S.® Bulk - Pipette Tips
Supplier: EPPENDORF
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Accessories for accu-jet® pro
Supplier: Brand
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
VWR®, Accessories for Mechanical Pipettes
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Supplier: HIRSCHMANN
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals