ContactUsLinkComponent
quick- disconnect coupler M16x1 Temperature range: -45°C…220°C 1 * 1 items
Catalog # HUBE10786
Supplier:  HUBER
quick- disconnect coupler M16x1 Temperature range: -45°C…220°C 1 * 1 items
Catalog # HUBE10786
Supplier:  HUBER
Frequently Bought Together