ContactUsLinkComponent
THE TEST AND CALIBRATION WEIGHTS KIT INCLUDES WEIGHTS AND TESTING MATERIALS REQUIRED TO KEEP THE TMA 4000 PROPERLY CALIBRATED. 1 * 1 items
Catalog # PERCL8041801
Supplier:  PerkinElmer
THE TEST AND CALIBRATION WEIGHTS KIT INCLUDES WEIGHTS AND TESTING MATERIALS REQUIRED TO KEEP THE TMA 4000 PROPERLY CALIBRATED. 1 * 1 items
Catalog # PERCL8041801
Supplier:  PerkinElmer
Frequently Bought Together