ContactUsLinkComponent
CloneSelect Imager system includes integrated 16 bit cooled CCD camera and 4x objective, fully configured computer with dedicated im aging software (including loci count and monoclonality reporting) pre-installed. 1 year warranty covering parts and l 1 * 1 items
Catalog # MLDVCLONESELECTIMA
Supplier:  Molecular Devices
CloneSelect Imager system includes integrated 16 bit cooled CCD camera and 4x objective, fully configured computer with dedicated im aging software (including loci count and monoclonality reporting) pre-installed. 1 year warranty covering parts and l 1 * 1 items
Catalog # MLDVCLONESELECTIMA
Supplier:  Molecular Devices
Frequently Bought Together