Orders LinkContactUsLinkComponent
Human/mouse reference RNA
Human/mouse reference RNA
Quantitative PCR human reference total RNA, pool of 10 human cell lines providing broad gene coverage qualified for use in QPCR, 25 µg, 1
Catalog # AGLS750500
Supplier:   AGILENT
Human/mouse reference RNA
Catalog # AGLS750500
Supplier:  AGILENT
CAS Number:  
Human/mouse reference RNA
Quantitative PCR human reference total RNA, pool of 10 human cell lines providing broad gene coverage qualified for use in QPCR, 25 µg, 1
CAS Number:  
Price
308,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Total RNA which has been extracted from cell lines representing different human tissues, pooled (10 human) or pooled (11 mouse) and treated to remove DNA to undetectable levels.
  • Greater consistency than RNA references derived from tissue sources
  • Maximum representation of low, medium, and high abundant gene transcripts
  • Manufactured at industrial scale lot sizes and stringently quality controlled for minimised lot-to-lot variation
  • No detectable levels of genomic DNA
Typical uses include optimisation of primer/probe sets to ensure no cross reactivity, as a standard to relatively quantitate unknowns to an unchanging reference for immediate validation without wasting precious RNA, or to compare QPCR efficiencies across labs/platforms/experiments.
Caution: For research use only. Not for use in diagnostic procedures.
Specifications
Specifications
  • Pk:
    1
  • Description:
    Quantitative PCR human reference total RNA, pool of 10 human cell lines providing broad gene coverage qualified for use in QPCR
  • Pk Info:
    25 µg