ContactUsLinkComponent
Universal Cutting Mill PULVERISETTE 19 50-700 rpm, for 200-240 V/1, 50-60 Hz, 2350 Watt Variable 50 and ndash 700 rpm for powerful comminution for large quantities 1 * 1 items
Catalog # FRIT19.4000.00
Supplier:  FRITSCH
Universal Cutting Mill PULVERISETTE 19 50-700 rpm, for 200-240 V/1, 50-60 Hz, 2350 Watt Variable 50 and ndash 700 rpm for powerful comminution for large quantities 1 * 1 items
Catalog # FRIT19.4000.00
Supplier:  FRITSCH
Frequently Bought Together