Sodium tartrate dihydrate Certified Reference Material for Karl Fischer Titration 15.66% Aquastar 1 * 100 g
Catalog # 1.06664.0103
Supplier: Merck
Sodium tartrate dihydrate Certified Reference Material for Karl Fischer Titration 15.66% Aquastar 1 * 100 g
Catalog # 1.06664.0103
Supplier: Merck
Frequently Bought Together
Bottle-top dispensers, Dispensette® S
Supplier: Brand
Lactose Standard 5 % Aquastar 1 * 10 g
Supplier: Merck
Filter tips, uTIP™
Supplier: BIOTIX
Sulphuric acid 95,0 - 100,5% 95.0-100.5% Ph. Eur.
Supplier: VWR Chemicals
L(+)-Ascorbic acid 99.0-100.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Syringe filters, Minisart® NML
Supplier: Sartorius
VWR® Automatic Burettes According to Dr. Schilling
Supplier: VWR Collection
Water Reag. Ph. Eur. 1095501 ammonium-free
Supplier: VWR Chemicals
Glass microfibre filters without binder, grade GF/D, Whatman™
Supplier: Whatman products (Cytiva)
Toluene, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Diethylamine for synthesis
Supplier: Merck
Potassium hydroxide ≥85%, pellets ACS, Fluka™
Supplier: Honeywell Chemicals
Reagents for water determination by the Coulometric Titration Karl Fischer method, Hydranal™
Supplier: Honeywell Chemicals
Fast green FCF
Supplier: Thermo Fisher Scientific
Water standards for Karl Fischer titration, Hydranal™
Supplier: Honeywell Chemicals
Sodium octanoate, Sigma-Aldrich®
Supplier: Merck