ContactUsLinkComponent
Heating Circulator Unistat T305w HT Temperature range:      (15) 65 ... 300 °C Heating capacity: 6 kW Pressure pump max.:     45 l/min / 0.9 bar Pump connection:  M24x1,5 male  Power supply:     400V 3~N 50/60Hz 1 * 1 items
Catalog # HUBE1003.0034.01
Supplier:  HUBER
Heating Circulator Unistat T305w HT Temperature range:      (15) 65 ... 300 °C Heating capacity: 6 kW Pressure pump max.:     45 l/min / 0.9 bar Pump connection:  M24x1,5 male  Power supply:     400V 3~N 50/60Hz 1 * 1 items
Catalog # HUBE1003.0034.01
Supplier:  HUBER
Frequently Bought Together