ContactUsLinkComponent
Waterproof alarm thermometer with immersion probe G1710-WPT3 Probe (permanently connected): With immersion probe, Ø3 mm, handle and cable shorttime 250°C, permanent 230°C Measuring input: Pt1000 (2-wire) Cable: Cable length 1 m silicone cable Display 1 * 1 items
Catalog # GRSG481378
Supplier:  Greisinger
Waterproof alarm thermometer with immersion probe G1710-WPT3 Probe (permanently connected): With immersion probe, Ø3 mm, handle and cable shorttime 250°C, permanent 230°C Measuring input: Pt1000 (2-wire) Cable: Cable length 1 m silicone cable Display 1 * 1 items
Catalog # GRSG481378
Supplier:  Greisinger
Frequently Bought Together