Multiparameter AP-2000-D set, consisting of GPS Aquameter, Aquaprobe AP-2000-D with DO (optical), EC, pH, Redox, Sal, depth and temperature and 2 extra ports for optional sensors (ISE or optical), with 3 meter cable, accessories and fluids in rugged 1 * 1 ST
Catalog # EIJK1857
Supplier: EIJKELKAMP AGRISECTOR
Frequently Bought Together