Orders LinkContactUsLinkComponent
Objectives for REVOLVE 3M microscope
Objectives for REVOLVE 3M microscope
Plan achromat objective, 10,6 mm
Catalog # ECHLJMAG0108
Supplier:   DISCOVER ECHO
Objectives for REVOLVE 3M microscope
Catalog # ECHLJMAG0108
Supplier:  DISCOVER ECHO
Objectives for REVOLVE 3M microscope
Objectives for REVOLVE 3M microscope
Plan achromat objective, 10,6 mm
Price
546,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Documents
Specifications
  • Magnification:
    10×
  • Description:
    Plan achromat objective
  • Seq:
    10
  • Working distance:
    10,6 mm