ContactUsLinkComponent
Unistat 705w with Pilot ONE, natural refrigerant -75...250 °C ,,TAC, self-optimizing (True Adaptive Control) 425 x 400 x 720 mm 1 * 1 items
Catalog # HUBE1068.0006.01
Supplier:  HUBER
Unistat 705w with Pilot ONE, natural refrigerant -75...250 °C ,,TAC, self-optimizing (True Adaptive Control) 425 x 400 x 720 mm 1 * 1 items
Catalog # HUBE1068.0006.01
Supplier:  HUBER
Frequently Bought Together