Order Entry
Austria
Orders LinkContactUsLinkComponent
Ionisation buffer solutions (CRMs), for AAS and ICP, Fluka™

Ionisation buffer solutions (CRMs), for AAS and ICP, Fluka™

2% Cesium (Cs) in 1% HNO₃, Plastic bottle, 100 mL
Catalog # HONCE4300-100ML
Supplier:   Honeywell Chemicals
Ionisation buffer solutions (CRMs), for AAS and ICP, Fluka™
Catalog # HONCE4300-100ML
CAS Number:  
Ionisation buffer solutions (CRMs), for AAS and ICP, Fluka™

2% Cesium (Cs) in 1% HNO₃, Plastic bottle, 100 mL

CAS Number:  
Price
249,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB)

Delivery information: Supplied complete with a detailed Certificate of Analysis (CoA) in every delivery.

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    2% Cesium (Cs) in 1% HNO₃

Specifications