Order Entry
Austria
Orders LinkContactUsLinkComponent
Cooling systems, CoolSafe™ | Avantor

Cooling systems, CoolSafe™

CoolSafe™ chambers, aluminium, 4 places, 50 ml tubes
Artikelnr. DIVECHAM-7050
Anbieter:   DIVERSIFIED BIOTECH
Cooling systems, CoolSafe™
Artikelnr. DIVECHAM-7050
Cooling systems, CoolSafe™
Cooling systems, CoolSafe™

CoolSafe™ chambers, aluminium, 4 places, 50 ml tubes

Preis
587,00 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents

Polystyrene cool safe box, aluminium cooling chamber.

  • Durable
  • Temperature stable
  • Corrosion resistant

Cooling system designed for 0,2 ml tubes (strips), 96 and 384 well microplates, 0,5, 1,5 and 5,0 ml tubes. Cool bricks with different temperature zones 1 or −10 °C.

Spezifikationen
  • For:
    50 ml tubes
  • Description:
    CoolSafe™ chambers, aluminium, 4 places
  • Seq:
    10
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.