Vivaspin Turbo 15 Regenerated Cellulose membrane, 10,000 MWCO, 48/PK ultrafiltration centrifugal concentrators for protein, DNA, nan oparticle and virus concentration for 4-15 mL sample volumes 1 * 48 items
Catalog # 512-0754
Supplier: Sartorius
Frequently Bought Together