Orders LinkContactUsLinkComponent
Accupod solid samplers
Accupod solid samplers
Accupod sampler, non sterile, HDPE, 5 ml
Catalog # 300-0587
Supplier:   Sampling Systems
Accupod solid samplers
Catalog # 300-0587
Supplier:  Sampling Systems
Accupod solid samplers
Accupod solid samplers
Accupod sampler, non sterile, HDPE, 5 ml
Price
216,00 €
Log in for availability
1 * 50 ST
Product Details & Documents
The AccuPod solid sampler is ideal for unit dose sampling of powders and granules.
  • Suitable for sampling blend uniformity
  • Next generation in sampling technology
  • Designed to minimise sampling error
The AccuPod is perfect for use in drug development and is frequently used in sampling blend uniformity. Ideal for sampling free flowing powders and granules. Sample remains in the pod until needed.
Specifications
  • Volume:
    5 ml
  • Description:
    Accupod sampler, non sterile
  • Material:
    HDPE
  • Seq:
    03