Order Entry
Austria
Orders LinkContactUsLinkComponent
Ammonia solution

Ammonia solution

500 mL
Catalog # SPCMAM180-500MLGL
Supplier:   Spectrum Chemical
Ammonia solution
Catalog # SPCMAM180-500MLGL
Supplier:  Spectrum Chemical
CAS Number:  1336-21-6
Ammonia solution

500 mL

CAS Number:  1336-21-6
Price
167,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

Strong Ammonia Solution, NF has many uses such as in topical pharmaceutical solutions destined for treating insect bites. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol

Documents

Specifications

Specifications

  • Pk:
    500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.