Orders LinkContactUsLinkComponent
Objectives
Objectives
HI PLAN achromatic objective, 12 mm
Catalog # LEIM11506228
Supplier:   LEICA MICROSYSTEMS
Objectives
Catalog # LEIM11506228
Supplier:  LEICA MICROSYSTEMS
Objectives
HI PLAN achromatic objective, 12 mm
Price
133,22 €
Log in for availability
1 * 1 ST
Product Details & Documents
Plan achromatic and phase contrast objectives.
Specifications
  • Magnification:
    10×/0,25
  • Description:
    HI PLAN achromatic objective
  • Working distance:
    12 mm