Digital Solutions
Applications
Support
About
Austria
Orders Link
ContactUsLinkComponent
Microscope Objectives
Objectives
Objectives
HI PLAN achromatic objective, 18 mm
LEIM11506226
:
LEICA MICROSYSTEMS
Objectives
LEIM11506226
:
LEICA MICROSYSTEMS
Objectives
HI PLAN achromatic objective, 18 mm
98,98 €
1 * 1 ST
-
+
Plan achromatic and phase contrast objectives.
Magnification:
4×/0,10
Description:
HI PLAN achromatic objective
Working distance:
18 mm