ContactUsLinkComponent
Aluminium length divider. Length 398,5 mm, height 56 mm. For plastic drawer of models MED-520, LABO-520,FROSTER-LABO-530, FROSTER-LABEX-530 1 * 1 items
Catalog # KIRS254108
Supplier:  KIRSCH PHILIPP
Aluminium length divider. Length 398,5 mm, height 56 mm. For plastic drawer of models MED-520, LABO-520,FROSTER-LABO-530, FROSTER-LABEX-530 1 * 1 items
Catalog # KIRS254108
Supplier:  KIRSCH PHILIPP
Frequently Bought Together