ContactUsLinkComponent
MABSol® Biotinylated Human CD19, Fc Tag, primary amine labeling (CD9-H8259) is expressed from human HEK293 cells. It contains AA Pr o 20 - Lys 291 (Accession P15391-1). It is the biotinylated form of Human CD19, Fc Tag (Cat. No. CD9-H5259). null
Catalog # ACRBCD9-H8259-25UG
Supplier:  ACROBIOSYSTEMS
MABSol® Biotinylated Human CD19, Fc Tag, primary amine labeling (CD9-H8259) is expressed from human HEK293 cells. It contains AA Pr o 20 - Lys 291 (Accession P15391-1). It is the biotinylated form of Human CD19, Fc Tag (Cat. No. CD9-H5259). null
Catalog # ACRBCD9-H8259-25UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together