ContactUsLinkComponent
Mouse IL-17A, His Tag (CT8-M5240) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Ala 158 (Accession Q62386-1). 1 * 1 mg
Catalog # ACRBCT8-M5240-1MG
Supplier:  ACRO Biosystems
Mouse IL-17A, His Tag (CT8-M5240) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Ala 158 (Accession Q62386-1). 1 * 1 mg
Catalog # ACRBCT8-M5240-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together