ContactUsLinkComponent
Recombinat Protein,FITC-Labeled Human ErbB3 Her3 Protein, His Tag,FITC-Labeled Human ErbB3, His Tag (ER3-HF2H5) is expressed from hu man 293 cells (HEK293). It contains AA Ser 20 - Thr 643 (Accession P21860-1). It is the FITC labeled form of Human Er 1 * 25 µG
Catalog # ACRBER3-HF2H5-25UG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,FITC-Labeled Human ErbB3 Her3 Protein, His Tag,FITC-Labeled Human ErbB3, His Tag (ER3-HF2H5) is expressed from hu man 293 cells (HEK293). It contains AA Ser 20 - Thr 643 (Accession P21860-1). It is the FITC labeled form of Human Er 1 * 25 µG
Catalog # ACRBER3-HF2H5-25UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together