ContactUsLinkComponent
Recombinat Protein,Mouse Dkk-3 Protein, His Tag,Mouse Dkk-3, His Tag (DK3-M5226) is expressed from human 293 cells (HEK293). It cont ains AA Pro 23 - Ile 349 (Accession NP056629.1).This protein carries a polyhistidine tag at the C-terminus. The prote 1 * 1 mg
Catalog # ACRBDK3-M5226-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Mouse Dkk-3 Protein, His Tag,Mouse Dkk-3, His Tag (DK3-M5226) is expressed from human 293 cells (HEK293). It cont ains AA Pro 23 - Ile 349 (Accession NP056629.1).This protein carries a polyhistidine tag at the C-terminus. The prote 1 * 1 mg
Catalog # ACRBDK3-M5226-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together