ContactUsLinkComponent
Recombinat Protein,Human Properdin Protein, His Tag,Human Properdin, His Tag (PRN-H52H3) is expressed from human 293 cells (HEK293). It contains AA Asp 28 - Leu 469 (Accession P27918-1).This protein carries a polyhistidine tag at the C-terminus. The 1 * 1 mg
Catalog # ACRBPRN-H52H3-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human Properdin Protein, His Tag,Human Properdin, His Tag (PRN-H52H3) is expressed from human 293 cells (HEK293). It contains AA Asp 28 - Leu 469 (Accession P27918-1).This protein carries a polyhistidine tag at the C-terminus. The 1 * 1 mg
Catalog # ACRBPRN-H52H3-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together