ContactUsLinkComponent
Mouse Flt-3 Ligand, His Tag (FLL-M52H4) is expressed from human 293 cells (HEK293). It contains AA Gly 27 - Arg 188 (Accession P4977 2-1). 1 * 100 µG
Catalog # ACRBFLL-M52H4100UG
Supplier:  ACRO Biosystems
Mouse Flt-3 Ligand, His Tag (FLL-M52H4) is expressed from human 293 cells (HEK293). It contains AA Gly 27 - Arg 188 (Accession P4977 2-1). 1 * 100 µG
Catalog # ACRBFLL-M52H4100UG
Supplier:  ACRO Biosystems
Frequently Bought Together