ContactUsLinkComponent
Mouse EGF, Fc Tag (EGF-M5265) is expressed from human 293 cells (HEK293). It contains AA Asn 977 - Arg 1029 (Accession P01132-1). 1 * 100 µG
  ACRBEGF-M5265100UG
Mouse EGF, Fc Tag (EGF-M5265) is expressed from human 293 cells (HEK293). It contains AA Asn 977 - Arg 1029 (Accession P01132-1). 1 * 100 µG
  ACRBEGF-M5265100UG
Frequently Bought Together