ContactUsLinkComponent
Human TIM-3, Fc Tag (TM3-H5258) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Arg 200 (Accession Q8TDQ0-1). 1 * 100 µG
Catalog # ACRBTM3-H5258100UG
Supplier:  ACRO Biosystems
Human TIM-3, Fc Tag (TM3-H5258) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Arg 200 (Accession Q8TDQ0-1). 1 * 100 µG
Catalog # ACRBTM3-H5258100UG
Supplier:  ACRO Biosystems
Frequently Bought Together