ContactUsLinkComponent
Mouse CD32b, His Tag (SPR BLI verified) (CDB-M52H7) is expressed from human 293 cells (HEK293). It contains AA Thr 40 - Arg 217 (Acc ession NP001070657.1). 1 * 100 µG
Catalog # ACRBCDB-M52H7100UG
Supplier:  ACRO Biosystems
Mouse CD32b, His Tag (SPR BLI verified) (CDB-M52H7) is expressed from human 293 cells (HEK293). It contains AA Thr 40 - Arg 217 (Acc ession NP001070657.1). 1 * 100 µG
Catalog # ACRBCDB-M52H7100UG
Supplier:  ACRO Biosystems
Frequently Bought Together