ContactUsLinkComponent
Human VEGF-B, His Tag (VE6-H5225) is expressed from human 293 cells (HEK293). It contains AA Pro 22 - Ala 207 (Accession P49765-1). 1 * 50 µG
Catalog # ACRBVE6-H5225-50UG
Supplier:  ACRO Biosystems
Human VEGF-B, His Tag (VE6-H5225) is expressed from human 293 cells (HEK293). It contains AA Pro 22 - Ala 207 (Accession P49765-1). 1 * 50 µG
Catalog # ACRBVE6-H5225-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together