ContactUsLinkComponent
Biotinylated Human IL-2 R beta, His,Avitag (ILB-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Asp 239 ( Accession P14784-1). 1 * 25 µG
Catalog # ACRBILB-H82E3-25UG
Supplier:  ACRO Biosystems
Biotinylated Human IL-2 R beta, His,Avitag (ILB-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Asp 239 ( Accession P14784-1). 1 * 25 µG
Catalog # ACRBILB-H82E3-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together