EPA Method 525.2 PCB Congeners Mix, 8-1046 500 µg/mL in Acetone 1 * 1 mL
EHERGA09000340AC
EPA Method 525.2 PCB Congeners Mix, 8-1046 500 µg/mL in Acetone 1 * 1 mL
EHERGA09000340AC
Frequently Bought Together
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection