ContactUsLinkComponent
384515253 HWS stirrer, propeller type, with KPG 10, for NW 150, 4 arms in stainless steel V4A / PFA beschichtet, plane Ø 100 mm length 450 mm, connection knurled Customs No. 73269092 1 * 1 items
Catalog # HWSA384515253
Supplier:  HWS LABORTECHNIK
384515253 HWS stirrer, propeller type, with KPG 10, for NW 150, 4 arms in stainless steel V4A / PFA beschichtet, plane Ø 100 mm length 450 mm, connection knurled Customs No. 73269092 1 * 1 items
Catalog # HWSA384515253
Supplier:  HWS LABORTECHNIK
Frequently Bought Together