0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
L-Tyrosine 1 * 100 mg
EHERC17896005
:
EHRENSTORFER
L-Tyrosine 1 * 100 mg
EHERC17896005
:
EHRENSTORFER
Frequently Bought Together
Nitrate standard solution, 1000 mg/L NO₃<sup>-</sup> in Water (from NaNO₃) Certipur®, Supelco®
: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
: Merck
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
: Merck
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
: Merck
VWR®, Conductivity standard solutions
: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
: VWR Chemicals
Triethanolamine (trolamine) ≥97%, GPR RECTAPUR®
: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
: Merck