ContactUsLinkComponent
Microscope stand Axio Observer 7 materials (Zeiss) - 6-position objective nosepiece, HD DIC M27 mot. ACR - motorized focus drive (min. step width 10 nm) - prepared for motorized beam path deflection - TFT touchscreen - 1x tube lens to fit with option 1 * 1 ST
Catalog # ZEIS4310099904000
Supplier:  ZEISS
Microscope stand Axio Observer 7 materials (Zeiss) - 6-position objective nosepiece, HD DIC M27 mot. ACR - motorized focus drive (min. step width 10 nm) - prepared for motorized beam path deflection - TFT touchscreen - 1x tube lens to fit with option 1 * 1 ST
Catalog # ZEIS4310099904000
Supplier:  ZEISS
Frequently Bought Together