SPM BUFFER 1 * 1.200 mL
Catalog # OMEGSPM-1200
Supplier: OMEGA BIO-TEK
Frequently Bought Together
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Decontamination solution, RNase AWAY®
Supplier: Thermo Fisher Scientific
Eppendorf® DNA LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
Bacterial DNA kit, E.Z.N.A.®
Supplier: OMEGA BIO-TEK
GelRed® & GelGreen® Nucleic acid gel stains
Supplier: Biotium
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
E.Z.N.A.® Plasmid Mini kit I
Supplier: OMEGA BIO-TEK
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Fine dosage syringes, Injekt®-F Solo
Supplier: B. Braun
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Ligation Sequencing Kits
Supplier: Oxford Nanopore Technologies
ELUTION BUFFER 1L 1 * 1 L
Supplier: OMEGA BIO-TEK