Order Entry
Austria
Orders LinkContactUsLinkComponent
Sack Clips

Sack Clips

Sack clip, Polyamide, 100 mm, 250 mm
Catalog # SAMP8010A-1
Supplier:   Sampling Systems
Sack Clips
Catalog # SAMP8010A-1
Supplier:  Sampling Systems
Sack Clips
Sack Clips

Sack clip, Polyamide, 100 mm, 250 mm

Price
218,00 €
Log in for availability
1 * 10 ST
Product Details & Documents

Sack clips are designed for the easy closing and sealing of bags.

  • Enables quick and easy sealing of bags and sacks
  • Heavy-duty design
  • Available in a range of materials
  • Available from stock
  • Fast delivery

The plastic sack clip is perfect for closing and sealing a range of different bags and sacks. Simply insert the bag into the jaws of the clip and click shut.

Specifications
  • Description:
    Sack clip
  • Length:
    250 mm
  • Material:
    Polyamide
  • Seq:
    01
  • Sealing length:
    100 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.