15 ML MICROSPH. SIZE STDS. 20 µM 1 * 15 mL
Catalog # MSPP4220A
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
pH buffers, ready to use
Supplier: WTW
Columbianadin phyproof(R) Reference Substance 1 * 10 mg
Supplier: Merck
Ganciclovir, Reference Standards
Supplier: USP
MICROSPH.SIZE STDS 10 µM + CERTIFICAAT 1 * 15 mL
Supplier: MARKET SOURCE PART PROCESS
Sulphur standard solution, 1000 mg/L S in water standard for ICP
Supplier: CPACHEM
Buffer solutions in sachets, perpHect® Pouches, Orion™
Supplier: Thermo Orion
Copper standard solution, 1000 mg/L Cu in 5% nitric acid (from Cu), standard for AAS, Specpure®
Supplier: Thermo Fisher Scientific
N-FORMYLKYNURENINE 1 * 100 mg
Supplier: Merck
Fenthion-oxon 10 ng/µl 1 * 10 mL
Supplier: EHRENSTORFER
VWR®, COD calibration standards, Certified Reference Materials (CRMs)
Supplier: VWR Chemicals
Polystyrene, analytical standards for GPC
Supplier: Merck
Standards for ion chromatograpghy
Supplier: CPACHEM
VWR®, Mineral Oil Rotational viscosity standards
Supplier: VWR Chemicals
pH Standard Solutions, IUPAC, Certified
Supplier: Hach