Orders LinkContactUsLinkComponent
Fibre optic ring lights
Fibre optic ring lights
Ring light, continuous, with flexible length approx. 1000 mm, int.Ø 81 mm, ext.Ø 106 mm, 40 - 110 mm working distance
Catalog # SCOC157081
Fibre optic ring lights
Catalog # SCOC157081
Fibre optic ring lights
Fibre optic ring lights
Ring light, continuous, with flexible length approx. 1000 mm, int.Ø 81 mm, ext.Ø 106 mm, 40 - 110 mm working distance
Price
955,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Fibre optic ring lights are available as six point, annular, or darkfield versions with specific advantages. For all ring lights, adapters are available for a variety of objective series. Ring lights are used to provide intense, uniform, and shadow-free illumination.
  • Mounts directly to the objective using a thumb screw
  • Housing made of rugged, black, anodised aluminium
  • Fibre bundle protected by flexible metal PVC sheathing
  • High white light transmission thanks to the use of SCHOTT PURAVIS ® fibres
Annular ring lights offer absolute shadow-free illumination. Six point ring lights are a cost effective alternative to annular ring lights. Darkfield ring lights offer low angle illumination for shiny, reflective surfaces.
Ordering information: Cold light source must be ordered separately.
Delivery information: Supplied without cold light source.
Specifications
  • Description:
    Ring light, continuous, with flexible length approx. 1000 mm, int.Ø 81 mm, ext.Ø 106 mm, 40 - 110 mm working distance
  • Seq:
    02