Order Entry
Austria
Orders LinkContactUsLinkComponent
VWR® Traceable® Lollipop™, Precision Thermometer

VWR® Traceable® Lollipop™, Precision Thermometer

Thermometer with 2.7 m cable, –50...250 °C (–58...482 °F), ±0.5 °C (-20 to 200 °C), ±1 °C otherwise, 0.1 °C, 5×3/16" Ø
Catalog # 620-2723
Supplier:   VWR Collection
VWR® Traceable® Lollipop™, Precision Thermometer
Catalog # 620-2723
Supplier:  VWR Collection
VWR® Traceable® Lollipop™, Precision Thermometer
VWR® Traceable® Lollipop™, Precision Thermometer

Thermometer with 2.7 m cable, –50...250 °C (–58...482 °F), ±0.5 °C (-20 to 200 °C), ±1 °C otherwise, 0.1 °C, 5×3/16" Ø

Price
106,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Water-resistant/shockproof thermometer (IP 64) replaces hazardous mercury units and is ideal for wet labs, wash-down areas, outdoors and virtually all field applications.

  • Backlit, rotating ½" high LCD digits are easy to view
  • °F/°C switchable

Shockproof in drop tests up to 5 feet. Stainless-steel (SS316) probe is resistant to most laboratory chemicals. Wide range covers 99 percent of the lab’s routine temperature measurements. Solid-state construction eliminates dangers inherent in breakable glass and toxic mercury thermometers.

Optional stainless-steel probe with cable features suction cups which allow versatile placement of water-resistant probe.

 : Multi-point calibration on an individually-numbered Traceable® Certificate which assures accuracy from our ISO/IEC 17025:2005 (1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement.

 : Supplied with Traceable® Certificate, stainless-steel probe, probe cover with pocket clip, and battery.

Specifications
  • Range:
    –50...250 °C (–58...482 °F)
  • Resolution:
    0.1 °C
  • Description:
    Thermometer with 2.7 m cable
  • Seq:
    02
  • Accuracy:
    ±0.5 °C (-20 to 200 °C), ±1 °C otherwise
  • Probe:
    3/16" Ø
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.