ContactUsLinkComponent
Connect Z®ONCE, spike BT (Bag Tip), 40 cm PVC (TOTM , tubing 3,0/4,1 mm (ID/OD), slide clamp, ONCE-LOCK, FlowStop, (Old Number: 76.4 373) 1 * 10 items
Catalog # CODA76.4368
Supplier:  CODAN
Connect Z®ONCE, spike BT (Bag Tip), 40 cm PVC (TOTM , tubing 3,0/4,1 mm (ID/OD), slide clamp, ONCE-LOCK, FlowStop, (Old Number: 76.4 373) 1 * 10 items
Catalog # CODA76.4368
Supplier:  CODAN
Frequently Bought Together