Orders LinkContactUsLinkComponent
VWR®, Wire baskets
VWR®, Wire baskets
140×140×140 mm
  215-4202
VWR®, Wire baskets
  215-4202
VWR®, Wire baskets
VWR®, Wire baskets
140×140×140 mm
108,00 €
1 * 1 ST
These wire baskets are made from rust-proof, electrolytically polished stainless steel.
  • Very good chemical and temperature resistance
  • Spot-welded mesh 8×8×1 mm
  • Reinforced edges
Can be used for transport, storage or sterilisation of laboratory utensils.
  • W×D×H:
    140×140×140 mm