VWR®, Wire baskets
100×100×100 mm
Catalog # 215-4200
Supplier: VWR Collection
Jump to:
Product Details & Documents
These wire baskets are made from rust-proof, electrolytically polished stainless steel.
- Very good chemical and temperature resistance
- Spot-welded mesh 8×8×1 mm
- Reinforced edges
Can be used for transport, storage or sterilisation of laboratory utensils.
Documents
Specifications
- W×D×H:100×100×100 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



