with FDA certificate Ø50mm x L=101mm, thread M 50x1,5 Trapezoidal groove TK 40, 1xM6 lateral 1xM10 lateral 1 * 1 items
Catalog # HWSA3792001/GK-P
Supplier: HWS LABORTECHNIK
with FDA certificate Ø50mm x L=101mm, thread M 50x1,5 Trapezoidal groove TK 40, 1xM6 lateral 1xM10 lateral 1 * 1 items
Catalog # HWSA3792001/GK-P
Supplier: HWS LABORTECHNIK
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals