ContactUsLinkComponent
DNA ladder 100bp. Ready to load DNA marker contains 10 DNA fragments ranging from 100bp to 1000bp with Xylene Cyanol FF and Orange G tracking dyes. (for 80 to 100 loads) 1 * 1.000 µl
Catalog # EMBIM3117
Supplier:  EMBITEC
DNA ladder 100bp. Ready to load DNA marker contains 10 DNA fragments ranging from 100bp to 1000bp with Xylene Cyanol FF and Orange G tracking dyes. (for 80 to 100 loads) 1 * 1.000 µl
Catalog # EMBIM3117
Supplier:  EMBITEC
Frequently Bought Together