ContactUsLinkComponent
Microbalance - 200 g x 0,01 mg with motorized automatic draftshield CUBIS 1 * 1 items
Catalog # SARBMCA225SS00A
Supplier:  Sartorius Balances
Microbalance - 200 g x 0,01 mg with motorized automatic draftshield CUBIS 1 * 1 items
Catalog # SARBMCA225SS00A
Supplier:  Sartorius Balances
Frequently Bought Together