ContactUsLinkComponent
Immersion probe for liquids / gases Sensor element: Pt100 Accuracies: DIN class AA (1/3 DIN class B) Connection: Mini-DIN plug, 4 wi re Probe length: 150 mm Probe diameter: Ø3 mm Cable and length: 1 m PVC cable Measuring range: -50 ... +400 °C 1 * 1 items
Catalog # GRSG600378
Supplier:  Greisinger
Immersion probe for liquids / gases Sensor element: Pt100 Accuracies: DIN class AA (1/3 DIN class B) Connection: Mini-DIN plug, 4 wi re Probe length: 150 mm Probe diameter: Ø3 mm Cable and length: 1 m PVC cable Measuring range: -50 ... +400 °C 1 * 1 items
Catalog # GRSG600378
Supplier:  Greisinger
Frequently Bought Together